Assay Details
Gene Symbol: LOC100284174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157070.1 | NP_001150542.1 | 458 | 63 | ||
GenBank mRNA | EU967362.1 | - | 458 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
