Assay Details
Gene Symbol: LOC100284461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157356.1 | NP_001150828.1 | 675 | 58 | ||
GenBank mRNA | EU968458.1 | - | 675 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
