Assay Details
Gene Symbol: LOC100284616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157511.1 | NP_001150983.1 | 485 | 68 | ||
GenBank mRNA | EU969050.1 | - | 485 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
