Assay Details
Gene Symbol: LOC100285495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158387.1 | NP_001151859.1 | 1151 | 96 | ||
GenBank mRNA | EU972523.1 | - | 1151 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
