Assay Details
Gene Symbol: LOC100285521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158412.1 | NP_001151884.1 | 888 | 73 | ||
GenBank mRNA | EU972637.1 | - | 888 | 73 |
Gene Symbol: LOC100381699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT062025.1 | - | 229 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT062025.1 transcripts.
Set Membership: |
![]() ![]() |
