Assay Details
Gene Symbol: LOC100285521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158412.1 | NP_001151884.1 | 1083 | 93 | ||
GenBank mRNA | EU972637.1 | - | 1083 | 93 |
Gene Symbol: LOC100381699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT062025.1 | - | 424 | 93 |
