Assay Details
Gene Symbol: LOC100286310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159197.1 | NP_001152669.1 | 243 | 90 | ||
GenBank mRNA | BT054925.1 | - | 218 | 90 | ||
EU976710.1 | - | 243 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
