Assay Details
Gene Symbol: LOC100281080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153999.1 | NP_001147471.1 | 313 | 69 | ||
GenBank mRNA | EU955527.1 | - | 313 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
