Assay Details
| Gene Symbol: LOC100281080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153999.1 | NP_001147471.1 | 614 | 85 | ||
| GenBank mRNA | EU955527.1 | - | 614 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100281080| Gene Symbol: LOC100281080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153999.1 | NP_001147471.1 | 614 | 85 | ||
| GenBank mRNA | EU955527.1 | - | 614 | 85 | ||