Assay Details
Gene Symbol: LOC100281139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154058.1 | NP_001147530.1 | 770 | 96 | ||
GenBank mRNA | EU955716.1 | - | 770 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
