Assay Details
Gene Symbol: LOC100281587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154506.1 | NP_001147978.1 | 2249 | 83 | ||
GenBank mRNA | BT053799.1 | - | 2185 | 83 | ||
EU957193.1 | - | 2249 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
