Assay Details
Gene Symbol: cl12627_1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154996.1 | NP_001148468.1 | 983 | 96 | ||
GenBank mRNA | BT060587.1 | - | 983 | 96 | ||
EU959499.1 | - | 986 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
