Assay Details
Gene Symbol: cl12627_1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154996.1 | NP_001148468.1 | 1141 | 101 | ||
GenBank mRNA | BT060587.1 | - | 1141 | 101 | ||
EU959499.1 | - | 1144 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
