Assay Details
Gene Symbol: LOC100283223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156125.1 | NP_001149597.1 | 430 | 66 | ||
GenBank mRNA | BT043085.1 | - | 394 | 66 | ||
EU963883.1 | - | 430 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
