Assay Details
| Gene Symbol: LOC100284728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157623.1 | NP_001151095.1 | 1611 | 58 | ||
| GenBank mRNA | EU969487.1 | - | 1611 | 58 | ||
More Information
Set Membership: |
|

LOC100284728
Genomic Map