Assay Details
Gene Symbol: LOC100284904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157799.1 | NP_001151271.1 | 807 | 67 | ||
GenBank mRNA | EU970063.1 | - | 807 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
