Assay Details
| Gene Symbol: LOC100285496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158388.1 | NP_001151860.1 | 1056 | 80 | ||
| GenBank mRNA | EU972526.1 | - | 1056 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285496| Gene Symbol: LOC100285496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158388.1 | NP_001151860.1 | 1056 | 80 | ||
| GenBank mRNA | EU972526.1 | - | 1056 | 80 | ||