Assay Details
Gene Symbol: LOC100286189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159077.1 | NP_001152549.1 | 1888 | 78 | ||
GenBank mRNA | EU976053.1 | - | 1888 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
