Assay Details
| Gene Symbol: ltk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111412.1 | NP_001104882.1 | 227 | 63 | ||
| GenBank mRNA | AF023165.1 | - | 227 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ltk2| Gene Symbol: ltk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111412.1 | NP_001104882.1 | 227 | 63 | ||
| GenBank mRNA | AF023165.1 | - | 227 | 63 | ||
Set Membership: |
|