Assay Details
| Gene Symbol: gsh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111672.1 | NP_001105142.1 | 268 | 92 | ||
| GenBank mRNA | AJ302783.1 | - | 268 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gsh1| Gene Symbol: gsh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111672.1 | NP_001105142.1 | 268 | 92 | ||
| GenBank mRNA | AJ302783.1 | - | 268 | 92 | ||
Set Membership: |
|