Assay Details
Gene Symbol: LOC542127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111759.1 | NP_001105229.1 | 834 | 73 | ||
GenBank mRNA | AY303682.1 | - | 834 | 73 |
Gene Symbol: LOC100501069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195900.1 | NP_001182829.1 | 1041 | 73 | ||
GenBank mRNA | BT060851.1 | - | 673 | 73 | ||
BT061899.1 | - | 671 | 73 | |||
BT061986.1 | - | 945 | 73 | |||
BT084414.1 | - | 1041 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
