Assay Details
| Gene Symbol: LOC542127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111759.1 | NP_001105229.1 | 1011 | 90 | ||
| GenBank mRNA | AY303682.1 | - | 1011 | 90 | ||
| Gene Symbol: LOC100501069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195900.1 | NP_001182829.1 | 1218 | 90 | ||
| GenBank mRNA | BT060851.1 | - | 850 | 90 | ||
| BT061899.1 | - | 848 | 90 | |||
| BT061986.1 | - | 1122 | 90 | |||
| BT084414.1 | - | 1218 | 90 | |||
More Information
Set Membership: |
|

LOC542127
Genomic Map