Assay Details
| Gene Symbol: RPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111921.1 | NP_001105391.1 | 443 | 59 | ||
| GenBank mRNA | BT035428.1 | - | 467 | 59 | ||
| EU952519.1 | - | 495 | 59 | |||
| U64436.1 | - | 443 | 59 | |||
More Information
Set Membership: |
|

RPS8
Genomic Map