Assay Details
Gene Symbol: glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112417.1 | NP_001105887.1 | 543 | 67 | ||
GenBank mRNA | M24845.1 | - | 543 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
