Assay Details
Gene Symbol: lox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112506.1 | NP_001105976.1 | 667 | 54 | ||
GenBank mRNA | BT063943.1 | - | 663 | 54 | ||
BT067473.1 | - | 663 | 54 | |||
BT085507.1 | - | 655 | 54 | |||
DQ335764.1 | - | 667 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
