Assay Details
| Gene Symbol: kin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112600.1 | NP_001106070.1 | 280 | 68 | ||
| GenBank mRNA | AY104404.2 | - | 291 | 68 | ||
| U66241.1 | - | 280 | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin1| Gene Symbol: kin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112600.1 | NP_001106070.1 | 280 | 68 | ||
| GenBank mRNA | AY104404.2 | - | 291 | 68 | ||
| U66241.1 | - | 280 | 68 | |||