Assay Details
| Gene Symbol: kin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112600.1 | NP_001106070.1 | 479 | 53 | ||
| GenBank mRNA | AY104404.2 | - | 491 | 53 | ||
| U66241.1 | - | 479 | 53 | |||
More Information
Set Membership: |
|

kin1
Genomic Map