Assay Details
Gene Symbol: LOC100279306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152327.1 | NP_001145799.1 | 1160 | 79 | ||
GenBank mRNA | BT040130.1 | - | 402 | 79 | ||
BT041473.1 | - | 367 | 79 | |||
BT054005.1 | - | 1160 | 79 | |||
BT064336.1 | - | 379 | 79 | |||
BT067566.1 | - | 374 | 79 | |||
EU960451.1 | - | 404 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
