Assay Details
Gene Symbol: LOC100279306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152327.1 | NP_001145799.1 | 1902 | 86 | ||
GenBank mRNA | BT040130.1 | - | 1144 | 86 | ||
BT041473.1 | - | 1109 | 86 | |||
BT054005.1 | - | 1902 | 86 | |||
BT064336.1 | - | 1121 | 86 | |||
BT067566.1 | - | 1116 | 86 | |||
EU960451.1 | - | 1146 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
