Assay Details
Gene Symbol: LOC100279948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152898.1 | NP_001146370.1 | 996 | 87 | ||
GenBank mRNA | BT055196.1 | - | 996 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
