Assay Details
Gene Symbol: LOC100279948 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152898.1 | NP_001146370.1 | 1358 | 84 | ||
GenBank mRNA | BT055196.1 | - | 1358 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
