Assay Details
Gene Symbol: LOC100280229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153159.1 | NP_001146631.1 | 268 | 90 | ||
GenBank mRNA | BT055818.1 | - | 268 | 90 | ||
HQ858782.1 | - | 127 | 90 |
