Assay Details
Gene Symbol: LOC100281385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154303.1 | NP_001147775.1 | 1585 | 100 | ||
GenBank mRNA | EU956567.1 | - | 1585 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
