Assay Details
Gene Symbol: LOC100281390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154308.1 | NP_001147780.1 | 1951 | 72 | ||
GenBank mRNA | EU956577.1 | - | 1951 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
