Assay Details
| Gene Symbol: LOC100281406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154324.1 | NP_001147796.1 | 1044 | 54 | ||
| GenBank mRNA | EU956608.1 | - | 1044 | 54 | ||
More Information
Set Membership: |
|

LOC100281406
Genomic Map