Assay Details
| Gene Symbol: LOC100285688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158579.1 | NP_001152051.1 | 1458 | 81 | ||
| GenBank mRNA | EU973426.1 | - | 1458 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100285688| Gene Symbol: LOC100285688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158579.1 | NP_001152051.1 | 1458 | 81 | ||
| GenBank mRNA | EU973426.1 | - | 1458 | 81 | ||