Assay Details
Gene Symbol: LOC100285706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158597.1 | NP_001152069.1 | 235 | 95 | ||
GenBank mRNA | EU973510.1 | - | 235 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
