Assay Details
Gene Symbol: LOC100285799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158689.1 | NP_001152161.1 | 530 | 88 | ||
GenBank mRNA | EU974027.1 | - | 530 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
