Assay Details
Gene Symbol: LOC100286076 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158964.1 | NP_001152436.1 | 1561 | 61 | ||
GenBank mRNA | EU975492.1 | - | 1561 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
