Assay Details
| Gene Symbol: cl3929_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153189.1 | NP_001146661.1 | 1473 | 92 | ||
| GenBank mRNA | BT055877.1 | - | 1473 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cl3929_1| Gene Symbol: cl3929_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153189.1 | NP_001146661.1 | 1473 | 92 | ||
| GenBank mRNA | BT055877.1 | - | 1473 | 92 | ||
Set Membership: |
|