Assay Details
Gene Symbol: LOC100281311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154229.1 | NP_001147701.1 | 514 | 69 | ||
GenBank mRNA | EU956295.1 | - | 514 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
