Assay Details
Gene Symbol: LOC100281541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154460.1 | NP_001147932.1 | 939 | 120 | ||
GenBank mRNA | EU957045.1 | - | 939 | 120 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
