Assay Details
Gene Symbol: LOC100282053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154966.1 | NP_001148438.1 | 754 | 95 | ||
GenBank mRNA | BT087337.1 | - | 693 | 95 | ||
EU959350.1 | - | 754 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
