Assay Details
| Gene Symbol: LOC100282425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155336.1 | NP_001148808.1 | 642 | 71 | ||
| GenBank mRNA | BT068780.1 | - | 564 | 71 | ||
| EU960851.1 | - | 642 | 71 | |||
More Information
Set Membership: |
|

LOC100282425
Genomic Map