Assay Details
| Gene Symbol: LOC100282451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155362.1 | NP_001148834.1 | 545 | 67 | ||
| GenBank mRNA | BT042064.2 | - | 625 | 67 | ||
| EU960949.1 | - | 545 | 67 | |||
More Information
Set Membership: |
|

LOC100282451
Genomic Map