Assay Details
Gene Symbol: LOC100285466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158359.1 | NP_001151831.1 | 446 | 100 | ||
GenBank mRNA | EU972378.1 | - | 446 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
