Assay Details
Gene Symbol: LOC100285580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158471.1 | NP_001151943.1 | 926 | 101 | ||
GenBank mRNA | EU972983.1 | - | 926 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
