Assay Details
Gene Symbol: LOC100286097 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158985.1 | NP_001152457.1 | 705 | 116 | ||
GenBank mRNA | EU975603.1 | - | 705 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
