Assay Details
Gene Symbol: pco118508(516) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146969.1 | NP_001140441.1 | 545 | 102 | ||
GenBank mRNA | BT038841.1 | - | 545 | 102 | ||
EU962942.1 | - | 557 | 102 |
