Assay Details
| Gene Symbol: pco118508(516) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146969.1 | NP_001140441.1 | 822 | 92 | ||
| GenBank mRNA | BT038841.1 | - | 822 | 92 | ||
| EU962942.1 | - | 834 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

pco118508(516)| Gene Symbol: pco118508(516) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146969.1 | NP_001140441.1 | 822 | 92 | ||
| GenBank mRNA | BT038841.1 | - | 822 | 92 | ||
| EU962942.1 | - | 834 | 92 | |||